Invitrogen
Manufacturer Code:PA5113127
Catalog # PIPA5113127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Tetratricopeptide repeat protein 30B protein (261-600AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2510042P03Rik fleer IFT70 IFT70B novel tetratrico peptide repeat (TPR) containing protein Tcc30b tetratricopeptide repeat domain 30B Tetratricopeptide repeat protein 30B TPR repeat protein 30B TTC30B; Tetratricopeptide repeat protein 30B; TPR repeat protein 30B
Gene Aliases: fleer; IFT70; IFT70B; TTC30B
UniProt ID: (Human) Q8N4P2
Entrez Gene ID: (Human) 150737
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.