Invitrogen
Manufacturer Code:PA557294
Catalog # PIPA557294
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human TTC21B |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Intraflagellar transport 139 homolog; putative protein product of Nbla10696; Tetratricopeptide repeat protein 21B; TPR repeat protein 21B
Gene Aliases: ATD4; IFT139; IFT139B; JBTS11; KIAA1992; Nbla10696; NPHP12; SRTD4; THM1; TTC21B
UniProt ID: (Human) Q7Z4L5
Entrez Gene ID: (Human) 79809
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.