Novus Biologicals
Manufacturer Code:NBP298556R
Catalog # NB104503
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human TTC19 fragment. |
Conjugate | DyLight 550 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC138312 MGC19520 tetratricopeptide repeat domain 192010204O13Rik tetratricopeptide repeat protein 19 mitochondrial TPR repeat protein 19; Tetratricopeptide repeat protein 19, mitochondrial; TPR repeat protein 19
Gene Aliases: 2010204O13Rik; MC3DN2; TTC19
UniProt ID: (Human) Q6DKK2
Entrez Gene ID: (Human) 54902
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.