Novus Biologicals
Manufacturer Code:NBP297168B
Catalog # NB101176
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human TTC11 fragment. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGI-135 Fis1 FIS1 homolog fission 1 (mitochondrial outer membrane) homolog (S. cerevisiae) fission 1 (mitochondrial outer membrane) homolog (yeast) H_NH0132A01.6 hFis1 mitochondrial fission 1 protein tetratricopeptide repeat domain 11 Tetratricopeptide repeat protein 11 TTC11TPR repeat protein 11; FIS1 homolog; fission 1 (mitochondrial outer membrane) homolog; H_NH0132A01.6; hFis1; Mitochondrial fission 1 protein; mitochondrial fission molecule; tetratricopeptide repeat domain 11; Tetratricopeptide repeat protein 11; TPR repeat protein 11
Gene Aliases: CGI-135; FIS1; TTC11
UniProt ID: (Human) Q9Y3D6
Entrez Gene ID: (Human) 51024
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.