Invitrogen
Manufacturer Code:PA5101607
Catalog # PIPA5101607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TSTA3 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-5 epimerase/4-reductase; 3-5 epimerase/4-reductase AI256181 FX GDP-4-keto-6-deoxy-D-mannose epimerase-reductase GDP-4-keto-6-deoxy-D-mannose-35-epimerase-4-reductase GDP-fucose synthase GDP-L-fucose synthase GDP-L-fucose synthetase P35b Protein FX red cell NADP(H)-binding protein SDR4E1 short chain dehydrogenase/reductase family 4E member 1 Short-chain dehydrogenase/reductase family 4E member 1 testis tissue sperm-binding protein Li 45a tissue specific transplantation antigen 3 tissue specific transplantation antigen P35B transplantation antigen P35B Tsta3 Tstap35b Tum-P35B antigen; GDP-4-keto-6-deoxy-D-mannose epimerase-reductase; GDP-4-keto-6-deoxy-D-mannose-3,5-epimerase-4-reductase; GDP-fucose synthase; GDP-L-fucose synthase; GDP-L-fucose synthetase; Protein FX; Red cell NADP(H)-binding protein; short chain dehydrogenase/reductase family 4E, member 1; Short-chain dehydrogenase/reductase family 4E member 1; testis tissue sperm-binding protein Li 45a; tissue specific transplantation antigen 3; Transplantation antigen P35B; Tum-P35B antigen
Gene Aliases: AI256181; FX; GFUS; P35B; SDR4E1; TSTA3; Tstap35b
UniProt ID: (Human) Q13630, (Mouse) P23591
Entrez Gene ID: (Human) 7264, (Mouse) 22122, (Rat) 300036
Molecular Function: dehydratase epimerase/racemase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.