Novus Biologicals
Manufacturer Code:NBP299639AF750
Catalog # NB062190
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human TST. |
Conjugate | Alexa Fluor 750 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.8.1 EC 2.8.1.1 MGC19578 RDSRhodanese thiosulfate sulfurtransferase thiosulfate sulfurtransferase (rhodanese); Rhodanese; Thiosulfate sulfurtransferase; thiosulfate sulfurtransferase (rhodanese)
Gene Aliases: RDS; TST
UniProt ID: (Human) Q16762
Entrez Gene ID: (Human) 7263
Molecular Function:
transfer/carrier protein
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.