Proteintech
Manufacturer Code:145311AP150UL
Catalog # 50-172-9521
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TSSC4 Fusion Protein Ag6030 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein TSSC4; Protein TSSC4 TSSC4; Tumor-suppressing STF cDNA 4 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 4 protein
Gene Aliases: TSSC4
UniProt ID: (Human) Q9Y5U2
Entrez Gene ID: (Human) 10078
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.