Proteintech
Manufacturer Code:162631AP150UL
Catalog # 50-173-0837
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TSR2 Fusion Protein Ag9328 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DT1P1A10 TSR2 WGG1; Pre-rRNA-processing protein TSR2 homolog; TSR2, 20S rRNA accumulation, homolog; WGG motif containing 1
Gene Aliases: 2310007F12Rik; DBA14; DT1P1A10; TSR2; WGG1
UniProt ID: (Human) Q969E8, (Mouse) Q8C8T8
Entrez Gene ID: (Human) 90121, (Rat) 317418, (Mouse) 69499
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.