Proteintech
Manufacturer Code:168871AP150UL
Catalog # 50-173-1238
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TSR1 Fusion Protein Ag10394 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KIAA1401 TSR1; Pre-rRNA-processing protein TSR1 homolog; TSR1, 20S rRNA accumulation, homolog; TSR1, 20S rRNA accumulation, homolog (S. cerevisiae)
Gene Aliases: AU040765; AW550801; KIAA1401; mKIAA1401; TSR1
UniProt ID: (Human) Q2NL82, (Mouse) Q5SWD9
Entrez Gene ID: (Human) 55720, (Mouse) 104662, (Rat) 100360406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.