Invitrogen
Manufacturer Code:MA535596
Catalog # PIMA535596
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0308 |
Immunogen | A synthesized peptide derived from human PBR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: benzodiazepine peripheral binding site; benzodiazepine receptor, peripheral; BPBS BZRP DBI IBP MBR mDRC Mitochondrial benzodiazepine receptor OTTHUMP00000028891 PBR PBS Peripheral-type benzodiazepine receptor pk18 PKBS PTBR Ptbzr PTBZR02 RATPTBZR02 Translocator protein Tspo Tspo1; isoquinoline-binding protein; Mitochondrial benzodiazepine receptor; PBR; Peripheral-type benzodiazepine receptor; PKBS; Translocator protein; translocator protein (18kDa)
Gene Aliases: BPBS; BZRP; DBI; IBP; MBR; mDRC; PBR; PBS; pk18; PKBS; PTBR; TSPO; Tspo1
UniProt ID: (Human) P30536, (Mouse) Q541E3
Entrez Gene ID: (Human) 706, (Mouse) 12257
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.