Invitrogen
Manufacturer Code:PA519088
Catalog # PIPA519088
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (RDNSGRRGGSRLPE) corresponding to the C-terminus amino acids of Tspo |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Benzodiazepin receptor (peripheral); benzodiazepine peripheral binding site; benzodiazepine receptor, peripheral; isoquinoline-binding protein; Mitochondrial benzodiazepine receptor; PBR; PBR Tspo translocator protein Bzrp IBP benzodiazepine receptor peripheral isoquinoline-binding protein peripheral benzodiazepine receptor; Peripheral-type benzodiazepine receptor; PKBS; Translocator protein; translocator protein (18kDa)
Gene Aliases: BPBS; BZRP; DBI; IBP; MBR; mDRC; PBR; PBS; pk18; PKBS; PTBR; Ptbzr; PTBZR02; RATPTBZR02; TSPO; Tspo1
UniProt ID: (Human) P30536, (Rat) P16257, (Mouse) Q541E3
Entrez Gene ID: (Human) 706, (Rat) 24230, (Mouse) 12257
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.