R&D Systems
Manufacturer Code:FAB8405U100UG
Catalog # FAB8405U100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 545422 |
Immunogen | NS0 mouse myeloma cell line transfected with human TSPAN33 Met1-Tyr283 Accession # NP_848657 |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: hPen; hPen PEN Penumbra proerythroblast new membrane tetraspanin 33 tetraspanin-33 tspan-33; Penumbra; Proerythroblast new membrane; Tetraspanin-33; Tspan-33
Gene Aliases: PEN; PEN.; TSPAN-33; TSPAN33
UniProt ID: (Human) Q86UF1
Entrez Gene ID: (Human) 340348
Molecular Function: cell adhesion molecule membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.