Invitrogen
Manufacturer Code:PA580195
Catalog # PIPA580195
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human TSPAN12 recombit protein (Position: G111-R224). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Tetraspan NET-2; Tetraspanin-12; Tetraspanin-12 Tspan-12 Tetraspan NET-2 Transmembrane 4 superfamily member 12 TSPAN12 NET2 TM4SF12 UNQ774/PRO1568; Transmembrane 4 superfamily member 12; Tspan-12
Gene Aliases: 9030619E17; AI426782; AI663988; AW111457; EVR5; NET-2; NET2; TM4SF12; TSPAN12; UNQ774/PRO1568
UniProt ID: (Human) O95859, (Rat) Q569A2, (Mouse) Q8BKT6
Entrez Gene ID: (Human) 23554, (Rat) 362326, (Mouse) 269831
Molecular Function:
cell adhesion molecule
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.