R&D Systems
Manufacturer Code:FAB9186V100UG
Catalog # FAB9186V100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 819202 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human TSPAN1 Met1-Gln241 Accession # O60635 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: NET1 NET-1 tetraspan 1 Tetraspan NET-1 tetraspanin 1 Tetraspanin TM4-C tetraspanin-1 TM4C TM4SF TSPAN-1; tetraspan 1; Tetraspan NET-1; Tetraspanin TM4-C; Tetraspanin-1; Tspan-1
Gene Aliases: NET1; TM4C; TM4SF; TSPAN1
UniProt ID: (Human) O60635
Entrez Gene ID: (Human) 10103
Molecular Function: cell adhesion molecule membrane-bound signaling molecule receptor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.