Novus Biologicals
Manufacturer Code:NBP233867
Catalog # NBP233867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: TMAEHFLTLLVVPAIKKDYGSQEDFTQVWNTTMKGLKCCGFTNYTDFEDSPYFKENSAFPPFCCNDNVTNTANETCTKQKAHDQKVEGCFNQLLYDIRTNAVTV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NET1 NET-1 tetraspan 1 Tetraspan NET-1 tetraspanin 1 Tetraspanin TM4-C tetraspanin-1 TM4C TM4SF TSPAN-1; tetraspan 1; Tetraspan NET-1; Tetraspanin TM4-C; Tetraspanin-1; Tspan-1
Gene Aliases: NET1; TM4C; TM4SF; TSPAN1
UniProt ID: (Human) O60635
Entrez Gene ID: (Human) 10103
Molecular Function:
cell adhesion molecule
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.