Proteintech
Manufacturer Code:160581AP150UL
Catalog # 50-173-0683
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TSPAN1 Fusion Protein Ag8786 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9030418M05Rik NET 1 RP11 322N21.1 Tetraspan NET 1 tetraspanin 1 Tetraspanin TM4 C TSPAN 1 TSPAN1; tetraspan 1; Tetraspan NET-1; Tetraspanin TM4-C; Tetraspanin-1; Tspan-1
Gene Aliases: 9030418M05Rik; NET1; TM4C; TM4SF; TSPAN1
UniProt ID: (Human) O60635, (Mouse) Q99J59, (Rat) Q6AYR9
Entrez Gene ID: (Human) 10103, (Mouse) 66805, (Rat) 298436
Molecular Function:
cell adhesion molecule
membrane-bound signaling molecule
receptor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.