Invitrogen
Manufacturer Code:PA588461
Catalog # PIPA588461
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-290 of human TSNAX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Translin-associated factor X; Translin-associated protein X; translin-like protein
Gene Aliases: R74833; TRAX; TSNAX
UniProt ID: (Human) Q99598, (Rat) Q9JHB5, (Mouse) Q9QZE7
Entrez Gene ID: (Human) 7257, (Rat) 64028, (Mouse) 53424
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.