Invitrogen
Manufacturer Code:MA524182
Catalog # PIMA524182
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 462310 |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse Tsukushi/TSK Thr18-Leu354 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: E2IG4 LRRC54 TSK E2-induced gene 4 protein leucine rich repeat containing 54 leucine-rich repeat-containing protein 54 tsukushi homolog tsukushi small leucine rich proteoglycan tsukushin; leucine rich repeat containing 54; Leucine-rich repeat-containing protein 54; Tsukushi; tsukushin
Gene Aliases: 9530051K01Rik; E2ig4; Lrrc54; Tsk; Tsku
UniProt ID: (Mouse) Q8CBR6
Entrez Gene ID: (Mouse) 244152
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.