Proteintech
Manufacturer Code:123701AP150UL
Catalog # 50-172-7933
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TSKU Fusion Protein Ag3054 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: E2 induced gene 4 protein E2IG4 LRRC54 TSK TSKU Tsukushi tsukushin; E2-induced gene 4 protein; leucine rich repeat containing 54; Leucine-rich repeat-containing protein 54; Tsukushi; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushin
Gene Aliases: 9530051K01Rik; E2IG4; LRRC54; TSK; TSKU; UNQ850/PRO1788
UniProt ID: (Human) Q8WUA8, (Mouse) Q8CBR6
Entrez Gene ID: (Human) 25987, (Mouse) 244152
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.