Invitrogen
Manufacturer Code:PA534344
Catalog # PIPA534344
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid synthetic peptide near the amino terminus of human TSHZ3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: teashirt 3; Teashirt homolog 3; teashirt zinc finger family member 3; TSHZ3 Teashirt zinc finger homeobox 3 TSH3 ZNF537; Zinc finger protein 537
Gene Aliases: A630038G13Rik; KIAA1474; mKIAA1474; teashirt3; TSH3; TSHZ3; Zfp537; ZNF537
UniProt ID: (Human) Q63HK5, (Rat) D3ZKB9, (Mouse) Q8CGV9
Entrez Gene ID: (Human) 57616, (Rat) 308523, (Mouse) 243931
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.