Novus Biologicals
Manufacturer Code:NBP254395PEATT594
Catalog # N254395P594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | TSHRB/1404 |
Immunogen | Recombinant human TSHR B-Chain protein |
Conjugate | PE/ATTO 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: CHNG1 LGR3hTSHR-I MGC75129 seven transmembrane helix receptor thyroid stimulating hormone receptor Thyroid-stimulating hormone receptor thyrotropin receptor thyrotropin receptor-I hTSHR-I TSH-R; seven transmembrane helix receptor; thyroid stimulating hormone receptor, isoform 2; Thyroid-stimulating hormone receptor; Thyrotropin receptor; thyrotropin receptor-I, hTSHR-I; TSH-R
Gene Aliases: CHNG1; hTSHR-I; LGR3; TSHR
UniProt ID: (Human) P16473
Entrez Gene ID: (Human) 7253
Molecular Function: extracellular matrix protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.