Novus Biologicals
Manufacturer Code:NBP180244
Catalog # NBP180244
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human TSG101. Peptide sequence RSELLELIQIMIVIFGEEPPVFSRPTVSASYPPYTATGPPNTSYMPGMPS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ESCRT-I complex subunit TSG101; ESCRT-I complex subunit TSG101 TSG10 tumor susceptibility gene 10 tumor susceptibility gene 101 tumor susceptibility gene 101 protein tumor susceptibility protein VPS23; tumor susceptibility gene 10; tumor susceptibility gene 101; Tumor susceptibility gene 101 protein; tumor susceptibility protein
Gene Aliases: TSG10; TSG101; VPS23
UniProt ID: (Human) Q99816
Entrez Gene ID: (Human) 7251
Molecular Function: ligase ubiquitin-protein ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.