Invitrogen
Manufacturer Code:PA568648
Catalog # PIPA568648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human TSC22D2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: TSC22 domain family 2; TSC22 domain family protein 2; TSC22 domain family protein 2-like protein; TSC22 domain family, member 2; TSC22-related-inducible leucine zipper protein 4; TSC22D2 KIAA0669 TILZ4 TSC22 domain family protein 2 TSC22-related-inducible leucine zipper protein 4
Gene Aliases: 1810043J12Rik; 5530402M19Rik; cb869; id:ibd5097; KIAA0669; TILZ4; TILZ4a; TILZ4b; TILZ4c; TSC22D2; wu:fa55e04; zgc:92475
UniProt ID: (Human) O75157
Entrez Gene ID: (Horse) 100057563, (Bovine) 541277, (Rabbit) 100337913, (Human) 9819, (Dog) 477111, (Guinea pig) 100718147, (Rat) 499624, (Mouse) 72033, (Zebrafish) 378875
Molecular Function:
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.