Invitrogen
Manufacturer Code:PA578208
Catalog # PIPA578208
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encompassing a sequence within the C-terminus region of human Hamartin/TSC1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hamartin; tuberous sclerosis 1 LAM TSC; Tuberous sclerosis 1 protein; Tuberous sclerosis 1 protein homolog; tumor suppressor
Gene Aliases: KIAA0243; LAM; TSC; TSC1
UniProt ID: (Human) Q92574, (Mouse) Q9EP53
Entrez Gene ID: (Human) 7248, (Mouse) 64930
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.