Abnova
Manufacturer Code:PAB13145
Catalog # 89-116-392
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal region 15 amino acids of human TSC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C for three months. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TSC1.
For Research Use Only
Protein Aliases: Hamartin; KIAA0243; LAM; MGC86987; TSC; Tuberous sclerosis 1 protein; tumor suppressor
Gene Aliases: KIAA0243; LAM; TSC; TSC1
UniProt ID: (Human) Q92574
Entrez Gene ID: (Human) 7248
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.