Novus Biologicals
Manufacturer Code:NBP15651520UL
Catalog # NBP15651520
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TRUB2(TruB pseudouridine (psi) synthase homolog 2 (E. coli)) The peptide sequence was selected from the middle region of TRUB2. Peptide sequence MNKSPMLITGIRCLYFAPPEFLLEVQCMHETQKELRKLVHEIGLELKTTA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLONE24922 EC 5.4.99.- probable tRNA pseudouridine synthase 2 RP11-339B21.1 TruB pseudouridine (psi) synthase homolog 2 (E. coli); Mitochondrial mRNA pseudouridine synthase TRUB2; TruB pseudouridine (psi) synthase family member 2; TruB pseudouridine (psi) synthase homolog 2
Gene Aliases: CLONE24922; TRUB2
UniProt ID: (Human) O95900
Entrez Gene ID: (Human) 26995
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.