Proteintech
Manufacturer Code:198911AP150UL
Catalog # 50-173-2587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRUB2 Fusion Protein Ag13726 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CLONE24922 RP11 339B21.1 TRUB2; Mitochondrial mRNA pseudouridine synthase TRUB2; probable tRNA pseudouridine synthase 2; TruB pseudouridine (psi) synthase family member 2; TruB pseudouridine (psi) synthase homolog 2
Gene Aliases: CLONE24922; G430055L02Rik; TRUB2
UniProt ID: (Human) O95900, (Mouse) Q91WG3, (Rat) Q5XFW2
Entrez Gene ID: (Human) 26995, (Mouse) 227682, (Rat) 366012
Molecular Function:
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.