Proteintech
Manufacturer Code:125201AP150UL
Catalog # 50-172-8037
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRUB1 Fusion Protein Ag3213 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Probable tRNA pseudouridine synthase 1; PUS4 TRUB1; TruB pseudouridine (psi) synthase family member 1; TruB pseudouridine (psi) synthase homolog 1
Gene Aliases: 2610009I02Rik; 9030425C13Rik; AI448235; BB118943; C76870; PUS4; TRUB1
UniProt ID: (Human) Q8WWH5, (Mouse) Q8C0D0
Entrez Gene ID: (Human) 142940, (Mouse) 72133
Molecular Function:
isomerase
lyase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.