Invitrogen
Manufacturer Code:OST00096G
Catalog # PIOST00096G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rabbit |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t extracellular loop of rabbit TRPV6 conjugated to an immunogenic carrier protein was used as the immunogen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: Transient receptor potential cation channel subfamily V member 6 TrpV6 Epithelial calcium channel 2 ECaC2 Calcium transport protein 1 CaT1 Trpv6; TRPV6
Entrez Gene ID: (Rabbit) 100009565
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.