Invitrogen
Manufacturer Code:OST00022W
Catalog # PIOST00022W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 4th cytoplasmic loop of rat TRPV6 conjugated to an immunogenic carrier protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: ABP/ZF CAT1 CATL ECAC2 HSA277909 LP6728 ZFAB; Calcium transport protein 1; CaT1; ECaC2; epithelial apical membrane calcium transporter/channel CaT1; Epithelial calcium channel 2; osmosensitive transient receptor potential channel 3; Transient receptor potential cation channel subfamily V member 6; transient receptor potential cation channel, subfamily 5, member 6; TrpV6
Gene Aliases: CaT1; Ecac2; Otrpc3; Trpv6
UniProt ID: (Rat) Q9R186
Entrez Gene ID: (Rat) 114246
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.