Invitrogen
Manufacturer Code:PA5114478
Catalog # PIPA5114478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRPV4 antibody was raised against an 18 amino acid peptide near the center of human TRPV4. The immunogen is located within amino acids 380 - 430 of TRPV4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 0610033B08Rik BCYM3 CMT2C HMSN2C OSM9-like transient receptor potential channel 4 Osm-9-like TRP channel 4 osmosensitive transient receptor potential channel 4 Otrpc4 SMAL SPSMA SSQTL1 Transient receptor potential cation channel subfamily V member 4 transient receptor potential cation channel subfamily 5 member 4 transient receptor potential cation channel subfamily V member 4 transient receptor potential protein 12 Trp12 TrpV4 trpv4 channel Vanilloi vanilloid receptor-like channel 2 Vanilloid receptor-like protein 2 vanilloid receptor-related osmotically activated channel vanilloid receptor-related osmotically activated channel (Vroac) vanilloid receptor-related osmotically-activated channel Vrl2 VRL-2 Vroac VR-OAC wu:fp52e02; Osm-9-like TRP channel 4; OSM9-like transient receptor potential channel 4; osmosensitive transient receptor potential channel 4; OTRPC4; Transient receptor potential cation channel subfamily V member 4; transient receptor potential cation channel, subfamily V, member 4; Transient receptor potential protein 12; TRP12; TrpV4; Vanilloid receptor-like channel 2; Vanilloid receptor-like protein 2; vanilloid receptor-related osmotically activated channel; Vanilloid receptor-related osmotically-activated channel; VR-OAC; VRL-2
Gene Aliases: BCYM3; CMT2C; HMSN2C; OTRPC4; SMAL; SPSMA; SSQTL1; TRP12; TRPV4; VRL2; VROAC
UniProt ID: (Human) Q9HBA0
Entrez Gene ID: (Human) 59341
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.