Invitrogen
Manufacturer Code:OST00146W
Catalog # PIOST00146W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Chicken |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t cytoplasmic loop of chicken TRPV4 (VRL-2 VRL2 OTRPC4 Vanilloid receptor-like channel 2) conjugated to an immunogenic carrier protein was used as the immunogen |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Transient receptor potential cation channel subfamily V member 4; TrpV4; Vanilloid receptor-related osmotically activated channel; vanilloid receptor-related osmotically activated channel protein; VR-OAC; VRL-2 VRL2 OTRPC4 Vanilloid receptor-like channel 2
Gene Aliases: TRPV4; VR-OAC
UniProt ID: (Chicken) A0A1D5PXA5
Entrez Gene ID: (Chicken) 395427
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.