Invitrogen
Manufacturer Code:OSR00063W
Catalog # PIOSR00063W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 1t cytoplasmic domain of mouse TRPV4 conjugated to an immunogenic carrier protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Osm-9-like TRP channel 4; osmosensitive transient receptor potential channel 4; OTRPC4; Transient receptor potential cation channel subfamily V member 4; Transient receptor potential cation channel subfamily V member 4 TrpV4 osm-9-like TRP channel 4 OTRPC4 Vanilloid receptor-like channel 2 Vanilloi; Transient receptor potential protein 12; TRP12; TrpV4; trpv4 channel; Vanilloid receptor-like channel 2; Vanilloid receptor-like protein 2; Vanilloid receptor-related osmotically-activated channel; VR-OAC
Gene Aliases: 0610033B08Rik; OTRPC4; Trp12; Trpv4; VR-OAC; VRL-2; Vrl2; VROAC
UniProt ID: (Mouse) Q9EPK8
Entrez Gene ID: (Mouse) 63873
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.