Invitrogen
Manufacturer Code:PA577322
Catalog # PIPA577322
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)HQPSLDQPAIPSSKAT corresponding to amino acid residues 413-428 of rat TRPV2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GRC; growth factor regulated channel; Growth factor-regulated calcium channel; Osm-9-like TRP channel 2; OTRPC2; Stretch-activated channel 2B; Transient receptor potential cation channel subfamily V member 2; TRPV2; Vanilloid receptor-like protein 1; VRL-1
Gene Aliases: GRC; OTRPC2; Sac2b; Trpv2; VRL-1; Vrl1
UniProt ID: (Mouse) Q9WTR1, (Rat) Q9WUD2
Entrez Gene ID: (Mouse) 22368, (Rat) 29465
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.