Invitrogen
Manufacturer Code:PA534498
Catalog # PIPA534498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 16 amino acid peptide near the carboxy terminus of human TRPV1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Capsaicin receptor; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TrpV1; TRPV1 Transient receptor potential cation channel subfamily V member 1 capsaicin receptor osm-9-like TRP channel 1 vanilloid receptor subtype 1 OTRPC1; Vanilloid receptor 1; vanilloid receptor subtype 1
Gene Aliases: TRPV1; VR1
UniProt ID: (Human) Q8NER1
Entrez Gene ID: (Human) 7442
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.