Invitrogen
Manufacturer Code:PA118350
Catalog # 01-670-602
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues C-Y(531) F S H L K E Y V A S(541) of human capsaicin receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Capsaicin receptor; cation channel; Osm-9-like TRP channel 1; OTRPC1; Transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential vanilloid 1 supraoptic nucleus; TrpV1; Vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR1 Transient receptor potential cation channel subfamily V member 1 TrpV1 osm-9-like TRP channel 1 OTRPC1 Capsaicin Receptor Vanilloid receptor
Gene Aliases: OTRPC1; Trpv1; TRPV1_SON; TRPV1alpha; TRPV1beta; VR-1; VR.5'sv; Vr1; Vr1l; Vr1l1
UniProt ID: (Rat) O35433, (Mouse) Q704Y3
Entrez Gene ID: (Rat) 83810, (Mouse) 193034
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.