Invitrogen
Manufacturer Code:PA577337
Catalog # PIPA577337
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)GRRASETERLLTPN corresponding to amino acid residues 6-19 of mouse TRPLM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MG-2; ML1; Mucolipidin; mucolipidosis type IV protein; Mucolipin-1; Transient receptor potential channel mucolipin 1; Transient receptor potential-mucolipin 1; TRPML1
Gene Aliases: 2210015I05Rik; MCOLN1; MG-2; ML4; MLIV; MST080; MSTP080; mucolipidin; TRP-ML1; TRPM-L1; TRPML1
UniProt ID: (Human) Q9GZU1, (Mouse) Q99J21
Entrez Gene ID: (Human) 57192, (Mouse) 94178, (Rat) 288371
Molecular Function: ion channel transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.