Novus Biologicals
Manufacturer Code:NB200145
Catalog # NB200145
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides made to residues 278-292 RNQLEKYISERTIQD and the C-terminus sequence NDLKGLLKEIANKIK of the human trp-p8 protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 6; Long transient receptor potential channel 6 LTrpC-6 LTRPC6 MGC2849 short form of the TRPM8 cationic channel transient receptor potential cation channel subfamily M member 8 Transient receptor potential p8 transient receptor potential subfamily M member 8 trp-p8 TRPP8transient receptor potential cation channel subfamily M member 8; LTrpC-6; short form of the TRPM8 cationic channel; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; Transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; Transient receptor potential p8; transient receptor potential subfamily M member 8; Trp-p8
Gene Aliases: LTRPC6; TRPM8; TRPP8
UniProt ID: (Human) Q7Z2W7
Entrez Gene ID: (Human) 79054
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.