Abnova
Manufacturer Code:PAB4072
Catalog # 89-131-263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to internal region of human TRPM8. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of TRPM8.
For Research Use Only
Protein Aliases: Long transient receptor potential channel 6; LTrpC-6; LTRPC6; MGC2849; short form of the TRPM8 cationic channel; transient receptor melastatin 8 variant 1; transient receptor melastatin 8 variant 2; Transient receptor potential cation channel subfamily M member 8; transient receptor potential cation channel, subfamily M, member 8; Transient receptor potential p8; transient receptor potential subfamily M member 8; Trp-p8; TRPP8
Gene Aliases: LTRPC6; TRPM8; TRPP8
UniProt ID: (Human) Q7Z2W7
Entrez Gene ID: (Human) 79054
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.