Novus Biologicals
Manufacturer Code:NBP212906AF405
Catalog # NBP212906W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S74-25 |
Immunogen | Fusion protein amino acids 1817-1863 (C- terminus) of mouse TrpM7 |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CHAK CHAK1FLJ25718 Channel-kinase 1 EC 2.7.11.1 Long transient receptor potential channel 7 LTRPC ion channel family member 7 LTrpC7 LTrpC-7 LTRPC7FLJ20117 transient receptor potential cation channel subfamily M member 7 transient receptor potential cation channel subfamily M member 7 transient receptor potential-phospholipase C-interacting kinase TRP-PLIK; Channel-kinase 1; Long transient receptor potential channel 7; LTRPC ion channel family member 7; LTrpC-7; Transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential-phospholipase C-interacting kinase
Gene Aliases: ALSPDC; CHAK; CHAK1; LTrpC-7; LTRPC7; TRP-PLIK; TRPM7
UniProt ID: (Human) Q96QT4
Entrez Gene ID: (Human) 54822
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.