Invitrogen
Manufacturer Code:PA587618
Catalog # PIPA587618
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 756-855 of human TRPM7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Channel-kinase 1; Long transient receptor potential channel 7; LTrpC-7; Transient receptor potential cation channel subfamily M member 7; transient receptor potential M7; Transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-related protein, ChaK; TRP-PLIK
Gene Aliases: 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; CHAK; CHAK1; Ltpr7; LTrpC-7; Ltrpc7; Trpm7; TRPPLIK
UniProt ID: (Mouse) Q923J1
Entrez Gene ID: (Mouse) 58800
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.