Invitrogen
Manufacturer Code:PA528101
Catalog # PIPA528101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 1802 and 1865 of Human LTRPC7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CHAK CHAK1 LTRPC7 TRP-PLIK LTRPC ion channel family member 7 LTrpC-7 channel-kinase 1 long transient receptor potential channel 7 transient receptor potential cation channel subfamily M member 7 transient receptor potential-phospholipase C-interacting kinase; Channel-kinase 1; Long transient receptor potential channel 7; LTRPC ion channel family member 7; LTrpC-7; Transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential-phospholipase C-interacting kinase
Gene Aliases: ALSPDC; CHAK; CHAK1; LTrpC-7; LTRPC7; TRP-PLIK; TRPM7
UniProt ID: (Human) Q96QT4
Entrez Gene ID: (Human) 54822
Molecular Function:
ion channel
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.