Invitrogen
Manufacturer Code:PA5102835
Catalog # PIPA5102835
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TRMP7 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2310022G15Rik 4833414K03Rik 5033407O22Rik ALSPDC CHAK CHAK1 Channel kinase 1 Channel-kinase 1 FLJ20117 Long transient receptor potential channel 7 long transient receptor potential channel 7 {ECO:0000250 Ltpr7 LTRPC ion channel family member 7 Ltrpc7 LTrpC-7 LTrpC7 {ECO:0000250 PubMed:11161216} RGD:620053} tr Transient receptor potential cation channel subfamily M member 7 transient receptor potential cation channel subfamily M member 7 {ECO:0000312 transient receptor potential cation channel subfamily M member 7 transient receptor potential M7 transient receptor potential-phospholipase C-interacting kinase transient receptor potential-phospholipase C-interacting kinase {ECO:0000303 transient receptor potential-related protein ChaK TRMP7 TRP PLIK Trpm7 trpm7 {ECO:0000312 TRPPLIK TRP-PLIK TRP-PLIK {ECO:0000303 UniProtKB:Q96QT4}; Channel-kinase 1; Long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250|UniProtKB:Q96QT4}; LTRPC ion channel family member 7; LTrpC-7; LTrpC7 {ECO:0000250|UniProtKB:Q96QT4}; Transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312|RGD:620053}; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; Transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303|PubMed:11161216}; transient receptor potential-related protein, ChaK; TRP-PLIK; TRP-PLIK {ECO:0000303|PubMed:11161216}; trpm7 {ECO:0000312|RGD:620053}
Gene Aliases: 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Ltpr7; LTrpC-7; LTRPC7; TRP-PLIK; TRPM7; TRPPLIK
UniProt ID: (Human) Q96QT4, (Mouse) Q923J1
Entrez Gene ID: (Human) 54822, (Mouse) 58800, (Rat) 679906
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.