Invitrogen
Manufacturer Code:PA19127
Catalog # PIPA19127
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
peptide-ELISA (pep-ELISA) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues C-TKESESTNSVRLML from the C-terminus of TRPM7 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Channel-kinase 1; Long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250|UniProtKB:Q96QT4}; LTRPC ion channel family member 7; LTrpC-7; LTrpC7 {ECO:0000250|UniProtKB:Q96QT4}; Transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312|RGD:620053}; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; Transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303|PubMed:11161216}; transient receptor potential-related protein, ChaK; TRMP7 CHAK CHAK1 LTRPC7 transient receptor potential cation channel subfamily M member 7; TRP-PLIK; TRP-PLIK {ECO:0000303|PubMed:11161216}; trpm7 {ECO:0000312|RGD:620053}
Gene Aliases: 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Ltpr7; LTrpC-7; LTRPC7; TRP-PLIK; TRPM7; TRPPLIK
UniProt ID: (Human) Q96QT4, (Mouse) Q923J1
Entrez Gene ID: (Human) 54822, (Mouse) 58800, (Rat) 679906
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.