Abnova
Manufacturer Code:H00054822A01
Catalog # 89-003-154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | TRPM7 (NP_060142.2 777 a.a. ~ 855 a.a) partial recombinant protein with GST tag. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant TRPM7.
For Research Use Only
Protein Aliases: CHAK; CHAK1; Channel-kinase 1; FLJ20117; FLJ25718; Long transient receptor potential channel 7; LTRPC ion channel family member 7; LTrpC-7; LTRPC7; Transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential-phospholipase C-interacting kinase; TRP-PLIK
Gene Aliases: ALSPDC; CHAK; CHAK1; LTrpC-7; LTRPC7; TRP-PLIK; TRPM7
UniProt ID: (Human) Q96QT4
Entrez Gene ID: (Human) 54822
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.