Invitrogen
Manufacturer Code:PA541040
Catalog # PIPA541040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TRPM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 5; long transient receptor potential-related channel 5; LTrpC-5; MLSN1 and TRP-related; MLSN1- and TRP-related gene 1 protein; Transient receptor potential cation channel subfamily M member 5; Transient receptor potential cation channel subfamily M member 5 LTRPC5 MTR1; transient receptor potential cation channel, subfamily M, member 5; TRPM5
Gene Aliases: 9430099A16Rik; LTrpC-5; LTRPC5; MTR1; TRPM5
UniProt ID: (Human) Q9NZQ8, (Mouse) Q9JJH7
Entrez Gene ID: (Horse) 100060376, (Dog) 483668, (Human) 29850, (Mouse) 56843, (Rat) 365391
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.