Invitrogen
Manufacturer Code:OST00111W
Catalog # PIOST00111W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from n-terminal region of human TRPM5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 5; LTrpC-5; LTRPC5 MLSN1- and TRP-related gene 1 protein TRPM5 LTRPC5 MTR1 Transient receptor potential cation channel subfamily M member 5; MLSN1 and TRP-related; MLSN1- and TRP-related gene 1 protein; Transient receptor potential cation channel subfamily M member 5; transient receptor potential cation channel, subfamily M, member 5; TRPM5
Gene Aliases: LTRPC5; MTR1; TRPM5
UniProt ID: (Human) Q9NZQ8
Entrez Gene ID: (Human) 29850
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.