Novus Biologicals
Manufacturer Code:NBP294006100UL
Catalog # NB022577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1026-1165 of human TRPM5 (NP_055370.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Long transient receptor potential channel 5; Long transient receptor potential channel 5 LTrpC5 LTrpC-5 LTRPC5transient receptor potential cation channel subfamily M member 5 MLSN1- and TRP-related gene 1 protein MTR1MLSN1 and TRP-related transient receptor potential cation channel subfamily M member 5; LTrpC-5; MLSN1 and TRP-related; MLSN1- and TRP-related gene 1 protein; Transient receptor potential cation channel subfamily M member 5; transient receptor potential cation channel, subfamily M, member 5; TRPM5
Gene Aliases: LTRPC5; MTR1; TRPM5
UniProt ID: (Human) Q9NZQ8
Entrez Gene ID: (Human) 29850
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.