OriGene
Manufacturer Code:CF500370
Catalog # 50-165-9449
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C4 |
Immunogen | Full-length protein expressed in 293T cell transfected with human TRPM4 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated non-selective cation channel 1; hTRPM4; Long transient receptor potential channel 4; LTrpC-4; LTrpC4 PFHB1B TRPM4B hTRPM4; Melastatin-4; Transient receptor potential cation channel subfamily M member 4; transient receptor potential cation channel, subfamily M, member 4
Gene Aliases: hTRPM4; LTRPC4; PFHB1B; TRPM4; TRPM4B
UniProt ID: (Human) A2RU25
Entrez Gene ID: (Human) 54795
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.