Invitrogen
Manufacturer Code:PA577324
Catalog # PIPA577324
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide EKEQSWIPKIFKK(C) corresponding to amino acid residues 5-17 of human TRPM4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcium-activated non-selective cation channel 1; hTRPM4; Long transient receptor potential channel 4; LTrpC-4; LTrpC4; melastatin like 2 protein; Melastatin-4; Melastatin-like 2; MLS2s; Transient receptor potential cation channel subfamily M member 4; transient receptor potential cation channel, subfamily M, member 4; transient receptor potential ion channel melastatin subgroup member 4; TRPM4
Gene Aliases: 1110030C19Rik; AW047689; hTRPM4; LTrpC-4; LTRPC4; Mls2s; PFHB1B; TRPM4; TRPM4B
UniProt ID: (Human) A2RU25, (Mouse) Q7TN37, (Rat) Q9ESQ5
Entrez Gene ID: (Human) 54795, (Mouse) 68667, (Rat) 171143
Molecular Function: ion channel receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.